Assay Details
Target Gene Details
Entrez Gene ID: | 20877 |
Gene Name: | aurora kinase B |
Gene Aliases: |
AIM-1, AIRK2, AL022959, Aik2, Aim1, Ark2, AurB, IPL1, STK-1, Stk12, Stk5 |
Location: |
Chr.11:69045625-69051664 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aurkb | NM_011496.1 | NP_035626.1 | ||
| AK075951.1 | BAC36078.1 | |||
| AK132006.1 | BAE20935.1 | |||
| AK182803.1 | ||||
| AK184704.1 | ||||
| BC003261.1 | AAH03261.1 | |||
| D21099.1 | BAA04658.1 |
More Information
Set Membership: |
|

Genomic Map