Assay Details
Target Gene Details
Entrez Gene ID: | 192976 |
Gene Name: | leucine rich repeat containing 75A |
Gene Aliases: |
Fam211a |
Location: |
Chr.11:62604884-62648583 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrc75a | NM_198861.1 | NP_942561.1 | ||
| AK146714.1 | BAE27379.1 | |||
| AK186775.1 | ||||
| AK189423.1 | ||||
| BC046404.1 | AAH46404.1 | |||
| BC053455.1 | AAH53455.1 |

Genomic Map