Assay Details
Target Gene Details
Entrez Gene ID: | 107476 |
Gene Name: | acetyl-Coenzyme A carboxylase alpha |
Gene Aliases: |
A530025K05Rik, Acac, Acc1, Gm738 |
Location: |
Chr.11:84129677-84401651 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acaca | NM_133360.2 | NP_579938.2 | ||
| AY451393.1 | AAS13685.1 |

Genomic Map