Assay Details
Target Gene Details
Entrez Gene ID: | 216560 |
Gene Name: | WD repeat containing planar cell polarity effector |
Gene Aliases: |
AV249152 |
Location: |
Chr.11:21572281-21898687 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wdpcp | NM_145425.3 | NP_663400.2 | ||
| AF424701.1 | AAL24810.1 | |||
| AK043672.1 | BAC31614.1 | |||
| AK082370.1 | ||||
| AK083042.1 | BAC38742.1 | |||
| BC141166.1 |

Genomic Map