Assay Details
Target Gene Details
Entrez Gene ID: | 216560 |
Gene Name: | WD repeat containing planar cell polarity effector |
Gene Aliases: |
AV249152 |
Location: |
Chr.11:21572281-21898687 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wdpcp | NM_145425.3 | 9 | 708 | NP_663400.2 |
| AF424701.1 | 9 | 671 | AAL24810.1 | |
| AK043672.1 | 8 | 595 | BAC31614.1 | |
| AK082370.1 | 9 | 619 | ||
| AK083042.1 | 9 | 709 | BAC38742.1 | |
| BC056139.1 | 1 | 42 | AAH56139.1 | |
| BC141166.1 | 9 | 672 |

Genomic Map