Assay Details
Target Gene Details
Entrez Gene ID: | 66156 |
Gene Name: | anaphase promoting complex subunit 11 |
Gene Aliases: |
1110011I19Rik, R75218 |
Location: |
Chr.11:120598421-120608198 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anapc11 | NM_001038230.2 | NP_001033319.1 | ||
| NM_025389.4 | NP_079665.1 | |||
| AA269705.1 | ||||
| AK003244.1 | BAB22663.1 | |||
| AK003612.1 | BAB22890.1 | |||
| AK045408.1 | BAC32348.1 | |||
| AK136358.1 | BAE22944.1 | |||
| AK158224.1 | ||||
| BC023039.1 | AAH23039.1 | |||
| BY074756.1 | ||||
| BY285612.1 | ||||
| W61838.1 |
More Information
Set Membership: |
|

Genomic Map