Assay Details
Target Gene Details
Entrez Gene ID: | 237877 |
Gene Name: | ATPase family, AAA domain containing 5 |
Gene Aliases: |
C130052G03Rik, FRAG1, Gm17 |
Location: |
Chr.11:80089379-80135797 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atad5 | NM_001029856.2 | NP_001025027.1 | ||
| AY557610.1 | AAT52048.1 | |||
| BC141180.1 |
More Information
Set Membership: |
|

Genomic Map