Assay Details
Target Gene Details
Entrez Gene ID: | 217310 |
Gene Name: | HID1 domain containing |
Gene Aliases: |
C630004H02Rik |
Location: |
Chr.11:115347705-115367753 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hid1 | NM_175454.2 | NP_780663.1 | ||
| AB087506.1 | BAC67686.1 | |||
| AK044395.1 | BAC31902.1 | |||
| AK049861.1 | ||||
| AK138882.1 | BAE23809.1 | |||
| BC024617.1 | AAH24617.1 |

Genomic Map