Assay Details
Target Gene Details
Entrez Gene ID: | 58996 |
Gene Name: | Rho GTPase activating protein 23 |
Gene Aliases: |
A330041B18Rik |
Location: |
Chr.11:97404393-97502402 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap23 | NM_021493.2 | 14 | 2820 | NP_067468.2 |
| AK173187.1 | 5 | 466 | BAD32465.1 | |
| BI739912.1 | 6 | 575 |

Genomic Map