Assay Details
Target Gene Details
Entrez Gene ID: | 382543 |
Gene Name: | ankyrin-repeat and fibronectin type III domain containing 1 |
Gene Aliases: |
Gm1175 |
Location: |
Chr.11:89421012-89696936 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankfn1 | NM_001080933.1 | 14 | 1972 | NP_001074402.1 |

Genomic Map