Assay Details
Target Gene Details
Entrez Gene ID: | 72605 |
Gene Name: | carbonic anhydrase 10 |
Gene Aliases: |
2700029L05Rik, BB085816, Ca10 |
Location: |
Chr.11:93098142-93601751 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car10 | NM_028296.3 | NP_082572.1 | ||
| AB080741.1 | BAC11853.1 | |||
| AK012302.1 | BAB28150.1 | |||
| AK035758.1 | BAC29176.1 | |||
| AK036003.1 | ||||
| AK038512.1 | BAC30021.1 | |||
| AK038695.1 | ||||
| AK132222.1 | BAE21042.1 | |||
| AK162576.1 | BAE36973.1 | |||
| BC017606.1 | AAH17606.1 | |||
| BC066111.1 | AAH66111.1 |

Genomic Map