Assay Details
Target Gene Details
Entrez Gene ID: | 72605 |
Gene Name: | carbonic anhydrase 10 |
Gene Aliases: |
2700029L05Rik, BB085816, Ca10 |
Location: |
Chr.11:93098142-93601751 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car10 | NM_028296.3 | 4 | 1353 | NP_082572.1 |
| AB080741.1 | 5 | 1309 | BAC11853.1 | |
| AK035758.1 | 4 | 588 | BAC29176.1 | |
| AK038695.1 | 5 | 1233 | ||
| AK132222.1 | 4 | 1194 | BAE21042.1 | |
| AK162576.1 | 4 | 471 | BAE36973.1 | |
| BC017606.1 | 4 | 545 | AAH17606.1 | |
| BC066111.1 | 5 | 1237 | AAH66111.1 |

Genomic Map