Assay Details
Target Gene Details
Entrez Gene ID: | 320150 |
Gene Name: | zinc finger, DHHC domain containing 17 |
Gene Aliases: |
A230053P19Rik, BB187739, D130071N24Rik, Hip14 |
Location: |
Chr.10:110941779-111010066 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zdhhc17 | NM_172554.2 | 13 | 1407 | NP_766142.2 |
| AK039575.1 | 13 | 1408 | ||
| AK046891.1 | 13 | 1349 | BAC32912.1 | |
| AK122406.1 | 12 | 2352 | BAC65688.1 | |
| BC117788.1 | 13 | 1349 | ||
| BC117789.2 | 13 | 1349 |

Genomic Map