Assay Details
Target Gene Details
Entrez Gene ID: | 109212 |
Gene Name: | family with sequence similarity 64, member A |
Gene Aliases: |
2610008F03Rik, 6720460F02Rik, AI115087, CATS |
Location: |
Chr.11:72042439-72047371 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam64a | NM_144526.3 | NP_653109.2 | ||
| AK011345.1 | ||||
| AK049710.1 | BAC33887.1 | |||
| AK078429.1 | BAC37270.1 | |||
| BC020115.1 | AAH20115.1 | |||
| BC029115.1 | AAH29115.1 |

Genomic Map