Assay Details
Target Gene Details
Entrez Gene ID: | 319767 |
Gene Name: | ATPase, class V, type 10B |
Gene Aliases: |
5930426O13Rik, 9030605H24Rik |
Location: |
Chr.11:42939557-43262285 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp10b | NM_176999.3 | NP_795973.2 |
More Information
Set Membership: |
|

Genomic Map