Assay Details
Target Gene Details
Entrez Gene ID: | 12291 |
Gene Name: | calcium channel, voltage-dependent, T type, alpha 1G subunit |
Gene Aliases: |
Cav3.1d, [a]1G, a1G, alpha-1G, mKIAA1123 |
Location: |
Chr.11:94408391-94474390 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacna1g | NM_001112813.2 | NP_001106284.1 | ||
| NM_001177888.1 | NP_001171359.1 | |||
| NM_001177890.1 | NP_001171361.1 | |||
| NM_009783.3 | NP_033913.2 | |||
| AJ012569.1 | CAB40793.1 | |||
| AK129294.1 | BAC98104.1 | |||
| BC057399.1 | AAH57399.1 | |||
| DQ317412.1 | ABC47034.1 |
More Information
Set Membership: |
|

Genomic Map