Assay Details
Target Gene Details
Entrez Gene ID: | 170720 |
Gene Name: | caspase recruitment domain family, member 14 |
Gene Aliases: |
Bimp2, CARMA2 |
Location: |
Chr.11:119307096-119345375 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Card14 | NM_130886.3 | 15 | 2223 | NP_570956.1 |
| AF363457.1 | 15 | 2202 | AAK60137.1 | |
| AK028888.1 | 10 | 1209 | BAC26175.1 | |
| BC004692.1 | 12 | 1658 | AAH04692.1 | |
| BC029102.1 | 15 | 2223 | AAH29102.1 |

Genomic Map