Assay Details
Target Gene Details
Entrez Gene ID: | 22288 |
Gene Name: | utrophin |
Gene Aliases: |
AA589569, DRP, Dmdl |
Location: |
Chr.10:12382187-12869685 on Build GRCm38 |
Assay Gene Location: | Within Exon 63 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Utrn | NM_011682.4 | 63 | 9204 | NP_035812.3 |
| AK035043.1 | 5 | 520 | ||
| X83506.1 | 14 | 1983 | CAA58496.1 | |
| Y12229.1 | 63 | 9112 | CAA72912.1 |
More Information
Set Membership: |
|

Genomic Map