Assay Details
Target Gene Details
Entrez Gene ID: | 320150 |
Gene Name: | zinc finger, DHHC domain containing 17 |
Gene Aliases: |
A230053P19Rik, BB187739, D130071N24Rik, Hip14 |
Location: |
Chr.10:110941779-111010066 on Build GRCm38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zdhhc17 | NM_172554.2 | NP_766142.2 | ||
| AK038670.1 | BAC30090.2 | |||
| AK038681.1 | BAC30097.2 | |||
| AK039575.1 | ||||
| AK046891.1 | BAC32912.1 | |||
| AK122406.1 | BAC65688.1 | |||
| AK134967.1 | ||||
| AK162922.1 | BAE37117.1 | |||
| BC058772.1 | AAH58772.1 | |||
| BC117788.1 | ||||
| BC117789.2 |
More Information
Set Membership: |
|

Genomic Map