Assay Details
Target Gene Details
Entrez Gene ID: | 66713 |
Gene Name: | ARP2 actin-related protein 2 |
Gene Aliases: |
4921510D23Rik, AA409782, Arp2, D6Ertd746e |
Location: |
Chr.11:20062304-20112951 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Actr2 | NM_146243.2 | NP_666355.1 | ||
| AK032832.1 | BAC28048.1 | |||
| AK088216.1 | BAC40216.1 | |||
| AK163217.1 | BAE37239.1 | |||
| BC027799.1 | AAH27799.1 | |||
| BC047530.1 | AAH47530.1 |
More Information
Set Membership: |
|

Genomic Map