Assay Details
Target Gene Details
Entrez Gene ID: | 268445 |
Gene Name: | ankyrin repeat domain 13b |
Gene Aliases: |
AW124583, B930093C12Rik |
Location: |
Chr.11:77470485-77489678 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd13b | NM_172945.2 | NP_766533.2 | ||
| BC071189.1 | AAH71189.1 | |||
| BC151158.1 | ||||
| CD578820.1 |

Genomic Map