Assay Details
Target Gene Details
Entrez Gene ID: | 12291 |
Gene Name: | calcium channel, voltage-dependent, T type, alpha 1G subunit |
Gene Aliases: |
Cav3.1d, [a]1G, a1G, alpha-1G, mKIAA1123 |
Location: |
Chr.11:94408391-94474390 on Build GRCm38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cacna1g | NM_001112813.2 | 33 | 6408 | NP_001106284.1 |
| NM_001177888.1 | 33 | 6387 | NP_001171359.1 | |
| NM_001177890.1 | 33 | 6336 | NP_001171361.1 | |
| NM_009783.3 | 34 | 6477 | NP_033913.2 | |
| AF227530.1 | 4 | 696 | AAF60165.1 | |
| AJ012569.1 | 34 | 6004 | CAB40793.1 | |
| AK129294.1 | 24 | 3180 | BAC98104.1 | |
| AK142458.1 | 12 | 1572 | BAE25073.1 | |
| BC057399.1 | 33 | 6175 | AAH57399.1 | |
| DQ317412.1 | 33 | 5926 | ABC47034.1 |
More Information
Set Membership: |
|

Genomic Map