Assay Details
Target Gene Details
Entrez Gene ID: | 75572 |
Gene Name: | acylphosphatase 2, muscle type |
Gene Aliases: |
2310004B09Rik |
Location: |
Chr.11:30505992-30649396 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acyp2 | NM_029344.3 | NP_083620.1 | ||
| AK009134.1 | BAB26095.2 | |||
| AK046238.1 | BAC32649.1 | |||
| BC027642.1 | AAH27642.1 |
More Information
Set Membership: |
|

Genomic Map