Assay Details
Target Gene Details
Entrez Gene ID: | 108686 |
Gene Name: | coiled coil domain containing 88A |
Gene Aliases: |
A430106J12Rik, AI848406, Ape, C130096N06Rik, C330012F17Rik, D130005J21Rik, Girdin, Giv, Grdn, Hkrp1 |
Location: |
Chr.11:29373595-29510808 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 18 - Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc88a | NM_176841.4 | NP_789811.2 | ||
| AB087827.1 | BAD98263.1 | |||
| AK129310.1 | BAC98120.1 | |||
| CN458498.1 | ||||
| CN532842.1 |
More Information
Set Membership: |
|

Genomic Map