Assay Details
Target Gene Details
Entrez Gene ID: | 216766 |
Gene Name: | gem (nuclear organelle) associated protein 5 |
Gene Aliases: |
AA407055, AA407208, AI451603, BB194447, C330013N08 |
Location: |
Chr.11:58120001-58168555 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gemin5 | NM_001166669.1 | 4 | 714 | NP_001160141.1 |
| NM_001166670.1 | 4 | 714 | NP_001160142.1 | |
| NM_001166671.1 | 4 | 593 | NP_001160143.1 | |
| NM_172558.3 | 4 | 714 | NP_766146.2 | |
| AK049216.1 | 4 | 649 | BAC33614.1 | |
| AK131137.1 | 4 | 569 | BAD21387.1 | |
| AK143536.1 | 4 | 663 | BAE25423.1 | |
| BC137802.1 | 4 | 657 | ||
| BC144922.1 | 4 | 657 | ||
| BY742908.1 |

Genomic Map