Assay Details
Target Gene Details
Entrez Gene ID: | 264895 |
Gene Name: | acyl-CoA synthetase family member 2 |
Gene Aliases: |
- |
Location: |
Chr.11:94557102-94601891 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acsf2 | NM_153807.2 | NP_722502.1 | ||
| AK034209.1 | BAC28632.1 | |||
| AK153582.1 | BAE32109.1 | |||
| BC018371.1 | AAH18371.1 | |||
| BC063269.1 | AAH63269.1 |
Target Gene Details
Entrez Gene ID: | 12643 |
Gene Name: | chondroadherin |
Gene Aliases: |
SLRR4A |
Location: |
Chr.11:94565074-94569127 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chad | NM_007689.4 | NP_031715.1 | ||
| AK158297.1 | BAE34451.1 | |||
| AK160489.1 | BAE35820.1 | |||
| BC012672.1 | AAH12672.1 |
More Information
Set Membership: |
|

Genomic Map