Assay Details
Target Gene Details
Entrez Gene ID: | 70894 |
Gene Name: | EF-hand calcium binding domain 3 |
Gene Aliases: |
4921510J17Rik |
Location: |
Chr.11:104934702-105117537 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efcab3 | NM_001081046.1 | NP_001074515.1 | ||
| AK014861.1 | ||||
| AK162207.1 | ||||
| BC050801.1 | AAH50801.1 |
More Information
Set Membership: |
|

Genomic Map