Assay Details
Target Gene Details
Entrez Gene ID: | 14030 |
Gene Name: | Ewing sarcoma breakpoint region 1 |
Gene Aliases: |
Ews, Ewsh |
Location: |
Chr.11:5069687-5099099 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ewsr1 | NM_001283061.1 | NP_001269990.1 | ||
| NM_001283062.1 | NP_001269991.1 | |||
| NM_001283063.1 | NP_001269992.1 | |||
| NM_007968.3 | NP_031994.2 | |||
| AK019460.1 | BAB31732.1 | |||
| AK034755.1 | ||||
| AK049743.1 | ||||
| AK146791.1 | ||||
| AK147103.1 | BAE27676.1 | |||
| AK147909.1 | BAE28221.1 | |||
| AK150345.1 | BAE29484.1 | |||
| AK151625.1 | BAE30560.1 | |||
| AK153276.1 | BAE31862.1 | |||
| AK215271.1 | ||||
| AK217040.1 | ||||
| BC068226.1 | AAH68226.1 | |||
| X79233.1 | CAA55815.1 |
More Information
Set Membership: |
|

Genomic Map