Assay Details
Target Gene Details
Entrez Gene ID: | 217335 |
Gene Name: | Fas (TNFRSF6) binding factor 1 |
Gene Aliases: |
1110033G01Rik, 5930413D20, mKIAA1863 |
Location: |
Chr.11:116142285-116168270 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbf1 | NM_172571.3 | 12 | 1460 | NP_766159.3 |
| AF241249.1 | 10 | 744 | AAK28326.1 | |
| AK086991.1 | ||||
| BC137794.1 | 10 | 736 | ||
| BC144918.1 | 10 | 736 |

Genomic Map