Assay Details
Target Gene Details
Entrez Gene ID: | 14401 |
Gene Name: | gamma-aminobutyric acid (GABA) A receptor, subunit beta 2 |
Gene Aliases: |
AI834970, C030002O17Rik, C030021G16Rik, Gabrab2, Gabrb-2 |
Location: |
Chr.11:42419757-42632591 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gabrb2 | NM_008070.3 | NP_032096.1 | ||
| AK090279.1 | BAC41155.1 | |||
| BC145973.1 | ||||
| BC145975.1 | ||||
| GU086167.1 | ||||
| U14419.1 | AAA79974.1 |
More Information
Set Membership: |
|

Genomic Map