Assay Details
Target Gene Details
Entrez Gene ID: | 319634 |
Gene Name: | EF-hand calcium binding domain 5 |
Gene Aliases: |
4930563A03Rik |
Location: |
Chr.11:77089904-77199943 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efcab5 | NM_176965.3 | NP_795939.2 | ||
| AK029790.1 | BAC26618.1 | |||
| AK035698.1 | BAC29157.1 |
More Information
Set Membership: |
|

Genomic Map