Assay Details
Target Gene Details
Entrez Gene ID: | 14406 |
Gene Name: | gamma-aminobutyric acid (GABA) A receptor, subunit gamma 2 |
Gene Aliases: |
GABAA-R, Gabrg-2, gamma2 |
Location: |
Chr.11:41910190-42000840 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
More Information
Set Membership: |
|

Genomic Map