Assay Details
Target Gene Details
Entrez Gene ID: | 216516 |
Gene Name: | coiled-coil domain containing 157 |
Gene Aliases: |
4930562D19Rik, mKIAA1656 |
Location: |
Chr.11:4141123-4160293 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc157 | NM_001164620.1 | NP_001158092.1 | ||
| NM_177616.3 | NP_808284.2 | |||
| AK076934.1 | BAC36532.1 | |||
| AK133165.1 | BAE21539.1 | |||
| AK143216.1 | ||||
| AK220409.1 | BAD90260.1 | |||
| BC046392.1 | 1 | 336 | AAH46392.1 | |
| BC120737.1 | AAI20738.1 | |||
| BC120739.1 | AAI20740.1 |

Genomic Map