Assay Details
Target Gene Details
Entrez Gene ID: | 237806 |
Gene Name: | dynein, axonemal, heavy chain 9 |
Gene Aliases: |
9030022M04, A230091C01, C230051G16, D11Ertd686e, Dnahc9, mKIAA0357 |
Location: |
Chr.11:65831324-66169186 on Build GRCm38 |
Assay Gene Location: | Within Exon 46 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnah9 | NM_001099633.1 | 46 | 8860 | NP_001093103.1 |
More Information
Set Membership: |
|

Genomic Map