Assay Details
Target Gene Details
Entrez Gene ID: | 13643 |
Gene Name: | ephrin B3 |
Gene Aliases: |
EFL-6, ELF-3, Elk-L3, Epl8, LERK-8, NLERK-2 |
Location: |
Chr.11:69554092-69560595 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Efnb3 | NM_007911.5 | NP_031937.1 | ||
| AF025288.1 | AAC53537.1 | |||
| AK048305.1 | BAC33299.1 | |||
| BC052001.1 | AAH52001.1 | |||
| BC058617.1 | AAH58617.1 |
More Information
Set Membership: |
|

Genomic Map