Assay Details
Target Gene Details
Entrez Gene ID: | 72194 |
Gene Name: | F-box and leucine-rich repeat protein 20 |
Gene Aliases: |
2610511F20Rik, 4632423N09Rik, AI849362, AL117906, C86145, Fbl2, Scr, mKIAA4147 |
Location: |
Chr.11:98082554-98150440 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxl20 | NM_028149.1 | 14 | 1354 | NP_082425.1 |
| AK012109.1 | 14 | 1078 | BAB28039.1 | |
| AK144786.1 | 14 | 1328 | BAE26066.1 | |
| AK220232.1 | 14 | 1307 | BAD90157.1 | |
| EF649694.1 | 14 | 1101 |

Genomic Map