Assay Details
Target Gene Details
Entrez Gene ID: | 14635 |
Gene Name: | galactokinase 1 |
Gene Aliases: |
AA409894, GALK, Gk, Glk, Glk1 |
Location: |
Chr.11:116008357-116012719 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galk1 | NM_016905.2 | 2 | 340 | NP_058601.2 |
| AB027012.1 | 2 | 296 | BAA84705.1 | |
| AK010340.1 | 2 | 337 | BAB26864.1 | |
| AK011103.1 | 2 | 341 | BAB27400.1 | |
| AK045694.1 | 2 | 314 | BAC32460.1 | |
| AK088151.1 | 2 | 341 | BAC40174.1 | |
| AK168716.1 | 2 | 300 | BAE40558.1 | |
| BC016602.1 | 2 | 340 | AAH16602.1 | |
| BC050151.1 | 2 | 311 | AAH50151.1 |
More Information
Set Membership: |
|

Genomic Map