Assay Details
Target Gene Details
Entrez Gene ID: | 11443 |
Gene Name: | cholinergic receptor, nicotinic, beta polypeptide 1 (muscle) |
Gene Aliases: |
Achr-2, Acrb |
Location: |
Chr.11:69784036-69795974 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrnb1 | NM_009601.4 | 10 | 1348 | NP_033731.3 |
| AK087554.1 | 10 | 1327 | BAC39924.1 | |
| BC023318.1 | 10 | 1354 | AAH23318.1 | |
| M14537.1 | 10 | 1291 | AAA37154.1 |
More Information
Set Membership: |
|

Genomic Map