Assay Details
Target Gene Details
Entrez Gene ID: | 14635 |
Gene Name: | galactokinase 1 |
Gene Aliases: |
AA409894, GALK, Gk, Glk, Glk1 |
Location: |
Chr.11:116008357-116012719 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galk1 | NM_016905.2 | NP_058601.2 | ||
| AB027012.1 | BAA84705.1 | |||
| AK010340.1 | BAB26864.1 | |||
| AK011103.1 | BAB27400.1 | |||
| AK045694.1 | BAC32460.1 | |||
| AK088151.1 | BAC40174.1 | |||
| AK168716.1 | BAE40558.1 | |||
| BC016602.1 | AAH16602.1 | |||
| BC050151.1 | AAH50151.1 |
More Information
Set Membership: |
|

Genomic Map