Assay Details
Target Gene Details
Entrez Gene ID: | 14607 |
Gene Name: | gastric inhibitory polypeptide |
Gene Aliases: |
- |
Location: |
Chr.11:96024534-96031296 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gip | NM_008119.2 | NP_032145.2 | ||
| AK008308.1 | BAB25592.1 | |||
| BC104314.1 | AAI04315.1 | |||
| BC104315.1 | AAI04316.1 |

Genomic Map