Assay Details
Target Gene Details
Entrez Gene ID: | 11448 |
Gene Name: | cholinergic receptor, nicotinic, epsilon polypeptide |
Gene Aliases: |
AChrepsilon, Acre, nAChRE |
Location: |
Chr.11:70614883-70619835 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrne | NM_009603.1 | 11 | 1296 | NP_033733.1 |
| AK006411.1 | 2 | 188 | BAB24575.1 | |
| X55718.1 | 11 | 1296 | CAA39251.1 |
More Information
Set Membership: |
|

Genomic Map