Assay Details
Target Gene Details
Entrez Gene ID: | 217304 |
Gene Name: | CD300 antigen like family member B |
Gene Aliases: |
CLM-7, Clm7, Gm253, LMIR5 |
Location: |
Chr.11:114922779-114934386 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd300lb | NM_199221.2 | 1 | 88 | NP_954691.2 |
| AK154359.1 | 1 | 89 | BAE32536.1 | |
| AK169466.1 | 1 | 62 | BAE41190.1 | |
| AK170613.1 | 1 | 60 | BAE41912.1 | |
| AY996128.1 |
More Information
Set Membership: |
|

Genomic Map