Assay Details
Target Gene Details
Entrez Gene ID: | 217304 |
Gene Name: | CD300 antigen like family member B |
Gene Aliases: |
CLM-7, Clm7, Gm253, LMIR5 |
Location: |
Chr.11:114922779-114934386 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd300lb | NM_199221.2 | NP_954691.2 | ||
| AB300703.1 | ||||
| AK154359.1 | BAE32536.1 | |||
| AK169466.1 | BAE41190.1 | |||
| AK170613.1 | BAE41912.1 | |||
| AY457053.1 | AAR27944.1 | |||
| AY996128.1 | ||||
| BC107352.1 | AAI07353.1 |
More Information
Set Membership: |
|

Genomic Map