Assay Details
Target Gene Details
Entrez Gene ID: | 11448 |
Gene Name: | cholinergic receptor, nicotinic, epsilon polypeptide |
Gene Aliases: |
AChrepsilon, Acre, nAChRE |
Location: |
Chr.11:70614883-70619835 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chrne | NM_009603.1 | NP_033733.1 | ||
| X55718.1 | CAA39251.1 |
More Information
Set Membership: |
|

Genomic Map