Assay Details
Target Gene Details
Entrez Gene ID: | 217149 |
Gene Name: | CDGSH iron sulfur domain 3 |
Gene Aliases: |
Mel-13, Mel13 |
Location: |
Chr.11:97685504-97688629 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cisd3 | NM_001085500.2 | NP_001078969.2 | ||
| AA987107.1 | ||||
| BC147739.1 | ||||
| BC147742.1 | ||||
| BC157903.1 |

Genomic Map