Assay Details
Target Gene Details
Entrez Gene ID: | 216705 |
Gene Name: | clathrin interactor 1 |
Gene Aliases: |
AI642036, AW537616, C530049I24Rik, Enth, Epn4, F630206G17Rik, mKIAA0171 |
Location: |
Chr.11:45851930-45910625 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clint1 | NM_001045520.3 | NP_001038985.2 | ||
| AB093212.1 | BAC41396.1 | |||
| AK143544.1 | BAE25429.1 | |||
| AK147875.1 | BAE28195.1 | |||
| AK167454.1 | BAE39540.1 | |||
| AK168991.1 | BAE40791.1 | |||
| BC004080.1 | AAH04080.1 |

Genomic Map