Assay Details
Target Gene Details
Entrez Gene ID: | 76884 |
Gene Name: | cytoplasmic FMR1 interacting protein 2 |
Gene Aliases: |
1500004I01Rik, 6430511D02Rik, AA930218, AU022376, Pir121, mKIAA1168 |
Location: |
Chr.11:46193849-46312859 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 24 - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyfip2 | NM_001252459.1 | NP_001239388.1 | ||
| NM_001252460.1 | NP_001239389.1 | |||
| NM_133769.3 | NP_598530.2 | |||
| AB093288.2 | BAC41472.2 | |||
| AF334144.1 | AAK81821.1 | |||
| AK030397.1 | BAC26942.1 | |||
| AK147224.1 | BAE27779.1 | |||
| AK147586.1 | BAE28010.1 | |||
| AK147632.1 | BAE28036.1 | |||
| AK147971.1 | ||||
| BC056974.1 | AAH56974.1 |
More Information
Set Membership: |
|

Genomic Map