Assay Details
Target Gene Details
Entrez Gene ID: | 16204 |
Gene Name: | fatty acid binding protein 6, ileal (gastrotropin) |
Gene Aliases: |
GT, I-15P, I-BABP, ILBP, ILBP3, Illbp |
Location: |
Chr.11:43596040-43601562 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fabp6 | NM_008375.2 | 2 | 209 | NP_032401.1 |
| BC119289.1 | 2 | 179 | ||
| BC120767.1 | 2 | 179 | ||
| CJ043022.1 | 2 | 209 |

Genomic Map