Assay Details
Target Gene Details
Entrez Gene ID: | 328035 |
Gene Name: | fatty acid desaturase domain family, member 6 |
Gene Aliases: |
BC044804, OTTMUSG00000021749 |
Location: |
Chr.11:115279622-115297822 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fads6 | NM_178035.4 | 3 | 541 | NP_828874.3 |
| AB087503.1 | 3 | 509 | BAC67683.1 | |
| BC044804.1 | 3 | 399 | AAH44804.1 | |
| BC132041.1 | 3 | 492 | ||
| BC132067.1 | 3 | 492 |

Genomic Map