Assay Details
Target Gene Details
Entrez Gene ID: | 76380 |
Gene Name: | centrosomal protein 112 |
Gene Aliases: |
1700001M19Rik, 1700029K01Rik, 8430407H02Rik, AV043680, AV207351, Ccdc46, Macoco |
Location: |
Chr.11:108425068-108860615 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cep112 | NM_029606.3 | NP_083882.3 | ||
| BC086808.1 | AAH86808.1 |

Genomic Map